Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyl-4-formylpiperidine-1-carboxylate 97%

Supplier: AMBEED, INC
Warning

A1442076-25G A1442076-5G
76661-026EA 35 USD InStock
76661-026 76661-028
Benzyl-4-formylpiperidine-1-carboxylate 97%
Benzyl-4-formylpiperidine-1-carboxylate
Precaución: For research use only.


Formula: C₁₄H₁₇NO₃
MDL Number: MFCD01317806
CAS Number: 138163-08-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty