Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hydroxymethanesulfinic acid 97%

Supplier: AMBEED, INC
Danger

A1442208-1000G A1442208-100G A1442208-500G
76661-050EA 19 USD InStock
76661-050 76661-052 76661-054
Hydroxymethanesulfinic acid 97%
Hydroxymethanesulfinic acid
Precaución: for Research Use Only


Formula: CH₄O₃S
MDL Number: MFCD00128934
CAS Number: 79-25-4
UN: 3261
ADR: 8,Ⅲ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty