Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[1,1'-Biphenyl]-4-carbonyl fluoride 97%

Supplier: AMBEED, INC
Danger

A1443042-1G A1443042-250MG A1443042-25G A1443042-5G
76661-084EA 132 USD InStock
76661-084 76661-086 76661-088 76661-090
[1,1'-Biphenyl]-4-carbonyl fluoride 97%
[1,1'-Biphenyl]-4-carbonyl fluoride
Precaución: for Research Use Only


Formula: C₁₃H₉FO
MW: 200.2084 g/mol
MDL Number: MFCD29066211
CAS Number: 2714-87-6
UN: 3261
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty