Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-3,6-dimethylbenzonitrile 97%

Supplier: AMBEED, INC
Danger

A1443316-100MG A1443316-1G A1443316-250MG
76661-138EA 151 USD InStock
76661-138 76661-140 76661-142
2-Fluoro-3,6-dimethylbenzonitrile 97%
2-Fluoro-3,6-dimethylbenzonitrile
Precaución: for Research Use Only


Formula: C₉H₈FN
MDL Number: MFCD28794319
CAS Number: 1805065-06-8
UN: 3276
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty