Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl 99% +(99.5%ee)

Supplier: AMBEED, INC
Warning

A1443764-100G A1443764-1G A1443764-5G
76661-216EA 228 USD InStock
76661-216 76661-218 76661-220
(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl 99% +(99.5%ee)
(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
Precaución: for Research Use Only


Formula: C₄₄H₃₂P₂
MDL Number: MFCD00010805
CAS Number: 76189-56-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty