Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 7-hydroxyheptanoate 98%

Supplier: AMBEED, INC
Warning

A1445860-100MG A1445860-250MG A1445860-5G
76661-300EA 20 USD InStock
76661-300 76661-302 76661-304
tert-Butyl 7-hydroxyheptanoate 98%
tert-Butyl 7-hydroxyheptanoate
Precaución: for Research Use Only


Formula: C₁₁H₂₂O₃
MDL Number: MFCD32706536
CAS Number: 86013-78-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty