Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromotriphenylene 95%

Supplier: AMBEED, INC
Warning

A1452239-1G A1452239-25G A1452239-5G A1452239-100G
76661-442EA 10 USD InStock
76661-442 76661-444 76661-446 77622-158
2-Bromotriphenylene 95%
2-Bromotriphenylene
Precaución: for Research Use Only


Formula: C₁₈H₁₁Br
MW: 307.19 g/mol
MDL Number: MFCD16659043
CAS Number: 19111-87-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty