Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxy-3-(trifluoromethoxy)benzamide 97%

Supplier: AMBEED, INC
Warning

A1452253-100MG A1452253-1G A1452253-250MG
76661-448EA 15 USD InStock
76661-448 76661-450 76661-452
2-Hydroxy-3-(trifluoromethoxy)benzamide 97%
2-Hydroxy-3-(trifluoromethoxy)benzamide
Precaución: for Research Use Only


Formula: C₈H₆F₃NO₃
MDL Number: MFCD18397431
CAS Number: 1261610-87-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty