Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diallyldimethylammonium chloride 60% in water

Supplier: AMBEED, INC
Warning

A1452309-1000G A1452309-100G A1452309-25G A1452309-500G
76661-462EA 20 USD InStock
76661-462 76661-464 76661-466 76661-468
Diallyldimethylammonium chloride 60% in water
Diallyldimethylammonium chloride
Precaución: for Research Use Only


Formula: C₈H₁₆ClN
MDL Number: MFCD00043200
CAS Number: 7398-69-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty