Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(tert-Butyl)phenyl trifluoromethanesulfonate 97%

Supplier: AMBEED, INC
Danger

A1453029-1G A1453029-5G A1453029-25G
76661-536EA 15 USD InStock
76661-536 76661-538 77622-160
4-(tert-Butyl)phenyl trifluoromethanesulfonate 97%
4-(tert-Butyl)phenyl trifluoromethanesulfonate
Precaución: for Research Use Only


Formula: C₁₁H₁₃F₃O₃S
MDL Number: MFCD28973056
CAS Number: 154318-75-9
UN: 1759
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty