Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl palmitoleate 98%

Supplier: AMBEED, INC
Warning

A1453697-1G A1453697-5G
76661-598EA 15 USD InStock
76661-598 76661-600
Methyl palmitoleate 98%
Methyl palmitoleate
Precaución: for Research Use Only


Formula: C₁₇H₃₂O₂
MDL Number: MFCD00042911
CAS Number: 1120-25-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty