Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl cis-13-docosenoate 95% (by GC)

Supplier: AMBEED, INC
Danger

A1453701-1G A1453701-25G A1453701-5G
76661-602EA 11 USD InStock
76661-602 76661-604 76661-606
Methyl cis-13-docosenoate 95% (by GC)
Methyl cis-13-docosenoate
Precaución: for Research Use Only


Formula: C₂₃H₄₄O₂
MDL Number: MFCD00027343
CAS Number: 1120-34-9
UN: 3272
ADR: 3,Ⅲ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty