Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Behenic acid 95%, (Chunks or pellets or flakes)

Supplier: AMBEED, INC
Warning

A1453786-1000G A1453786-100G A1453786-25G A1453786-500G
76661-626EA 52 USD InStock
76661-626 76661-628 76661-630 76661-632
Behenic acid 95%, (Chunks or pellets or flakes)
Behenic acid
Precaución: for Research Use Only


Formula: C₂₂H₄₄O₂
MDL Number: MFCD00002807
CAS Number: 112-85-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty