Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dodecylbenzene 98% + (Branch chain mixture of isomers)

Supplier: AMBEED, INC
Warning

A1454213-100G A1454213-25G A1454213-5G
76661-658EA 124 USD InStock
76661-658 76661-660 76661-662
Dodecylbenzene 98% + (Branch chain mixture of isomers)
Dodecylbenzene
Precaución: for Research Use Only


Formula: C₁₈H₃₀
MW: 246.44 g/mol
Boiling Pt: 275
Melting Pt: -4
MDL Number: MFCD00008974
CAS Number: 123-01-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty