Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Docosyl acrylate ≥95.0% (by GC) stabilised

Supplier: AMBEED, INC
Warning

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A1455581-25G A1455581-5G
76661-726EA 17 USD InStock
76661-726 76661-728
Docosyl acrylate ≥95.0% (by GC) stabilised
Docosyl acrylate
Precaución: for Research Use Only


Formula: C₂₅H₄₈O₂
MDL Number: MFCD00080409
CAS Number: 18299-85-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty