Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Crocein orange G BS

Supplier: AMBEED, INC
Warning

A1455667-100G A1455667-25G A1455667-500G
76661-734EA 60 USD InStock
76661-734 76661-736 76661-738
Crocein orange G BS
Crocein orange G
Precaución: for Research Use Only


Formula: C₁₆H₁₂N₂O₄S
MW: 350.32 g/mol
MDL Number: MFCD00003912
CAS Number: 1934-20-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty