Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Isobutyl-3-methoxypyrazine 99+%

Supplier: AMBEED, INC
Warning

A1456027-100G A1456027-1G A1456027-25G A1456027-5G
76661-758EA 299 USD InStock
76661-758 76661-760 76661-762 77624-208
2-Isobutyl-3-methoxypyrazine 99+%
2-Isobutyl-3-methoxypyrazine
Precaución: for Research Use Only


Formula: C₉H₁₄N₂O
MDL Number: MFCD00006128
CAS Number: 24683-00-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty