Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α-Linolenic acid methyl ester 80%

Supplier: AMBEED, INC
Warning

A1456329-25G A1456329-100G
76661-766EA 36 USD InStock
76661-766 77566-954
α-Linolenic acid methyl ester 80%
α-Linolenic acid methyl ester
Precaución: For research use only.


Formula: C₁₉H₃₂O₂
MDL Number: MFCD00135851
CAS Number: 301-00-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty