Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,18-Octadecanedioic acid 99%

Supplier: AMBEED, INC
Warning

A1459984-100G A1459984-1G A1459984-25G A1459984-500G A1459984-5G
76661-900EA 240 USD InStock
76661-900 76661-902 76661-904 76661-906 76661-908
1,18-Octadecanedioic acid 99%
1,18-Octadecanedioic acid
Precaución: for Research Use Only


Formula: C₁₈H₃₄O₄
MW: 314.47 g/mol
MDL Number: MFCD00142369
CAS Number: 871-70-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty