Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Aminoacetophenone 99%

Supplier: AMBEED, INC
Warning

A1460642-1000G A1460642-100G A1460642-25G A1460642-500G
76661-924EA 131 USD InStock
76661-924 76661-926 76661-928 76661-930
4'-Aminoacetophenone 99%
4'-Aminoacetophenone
Precaución: for Research Use Only


Formula: C₈H₉NO
MW: 135.17 g/mol
Boiling Pt: 293
MDL Number: MFCD00007896
CAS Number: 99-92-3
Merck Index: 13,00412
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty