Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Di(1H-pyrazol-3-yl)pyridine 97%

Supplier: AMBEED, INC
Warning

A1462360-1G A1462360-250MG A1462360-25G A1462360-5G
76662-062EA 92 USD InStock
76662-062 76662-064 76662-066 76662-068
2,6-Di(1H-pyrazol-3-yl)pyridine 97%
2,6-Di(1H-pyrazol-3-yl)pyridine
Precaución: for Research Use Only


Formula: C₁₁H₉N₅
MDL Number: MFCD29049605
CAS Number: 63285-53-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty