Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 2-(piperazin-1-yl)acetate hydrochloride 97%

Supplier: AMBEED, INC
Warning

A1462499-250MG A1462499-5G
76662-082EA 34 USD InStock
76662-082 76662-084
tert-Butyl 2-(piperazin-1-yl)acetate hydrochloride 97%
tert-Butyl 2-(piperazin-1-yl)acetate hydrochloride
Precaución: for Research Use Only


Formula: C₁₀H₂₁ClN₂O₂
CAS Number: 2408342-38-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty