Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Di-tert-butyl ((3-bromo-6-chloropyridin-2-yl)methyl)iminodicarbonate 97%

Supplier: AMBEED, INC
Warning

A1462810-100MG A1462810-1G A1462810-250MG A1462810-5G
76662-118EA 104 USD InStock
76662-118 76662-120 76662-122 76662-124
Di-tert-butyl ((3-bromo-6-chloropyridin-2-yl)methyl)iminodicarbonate 97%
Di-tert-butyl ((3-bromo-6-chloropyridin-2-yl)methyl)iminodicarbonate
Precaución: for Research Use Only


Formula: C₁₆H₂₂BrClN₂O₄
MDL Number: MFCD34186947
CAS Number: 2703752-57-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty