Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

rel-Potassium sodium (2R,3R)-2,3-dihydroxysuccinate tetrahydrate 99% (contains 20-30% H₂O)

Supplier: AMBEED, INC
Warning

A1464179-1000G A1464179-100G A1464179-500G
76662-278EA 301 USD InStock
76662-278 76662-280 76662-282
rel-Potassium sodium (2R,3R)-2,3-dihydroxysuccinate tetrahydrate 99% (contains 20-30% H₂O)
rel-Potassium sodium (2R,3R)-2,3-dihydroxysuccinate tetrahydrate
Precaución: for Research Use Only


Formula: C₄H₁₂KNaO₁₀
CAS Number: 57527-98-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty