Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nicotinonitrile 99+%

Supplier: AMBEED, INC
Warning

A1464560-1000G A1464560-100G A1464560-25G A1464560-500G
76662-336EA 103 USD InStock
76662-336 76662-338 76662-340 76662-342
Nicotinonitrile 99+%
Nicotinonitrile
Precaución: for Research Use Only


Formula: C₆H₄N₂
MDL Number: MFCD00087130
CAS Number: 100-54-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty