Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl (2-(2-(2-iodoethoxy)ethoxy)ethyl)carbamate 97%

Supplier: AMBEED, INC
Warning

A1464632-100MG A1464632-1G A1464632-250MG A1464632-5G
76662-352EA 13 USD InStock
76662-352 76662-354 76662-356 76662-358
tert-Butyl (2-(2-(2-iodoethoxy)ethoxy)ethyl)carbamate 97%
tert-Butyl (2-(2-(2-iodoethoxy)ethoxy)ethyl)carbamate
Precaución: for Research Use Only


Formula: C₁₁H₂₂INO₄
MDL Number: MFCD31580051
CAS Number: 1820026-89-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty