Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl adipoyl chloride 97%

Supplier: AMBEED, INC
Danger

A1466982-1G A1466982-25G A1466982-5G
76662-468EA 34 USD InStock
76662-468 76662-470 76662-472
Methyl adipoyl chloride 97%
Methyl adipoyl chloride
Precaución: for Research Use Only


Formula: C₇H₁₁ClO₃
MW: 178.62 g/mol
MDL Number: MFCD00013661
CAS Number: 35444-44-1
UN: 3265
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty