Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

12-Hydroxystearic acid 85+%

Supplier: AMBEED, INC
Warning

A1467042-100G A1467042-25G A1467042-500G
76662-492EA 9 USD InStock
76662-492 76662-494 76662-496 76662-498
12-Hydroxystearic acid 85+%
12-Hydroxystearic acid
Precaución: for Research Use Only


Formula: C₁₈H₃₆O₃
MW: 300.48 g/mol
MDL Number: MFCD00004592
CAS Number: 106-14-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty