Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl nitroacetate 98%

Supplier: AMBEED, INC
Warning

A1467780-100G A1467780-1G A1467780-25G A1467780-5G
76662-584EA 223 USD InStock
76662-584 76662-586 76662-588 76662-590
Methyl nitroacetate 98%
Methyl nitroacetate
Precaución: for Research Use Only


Formula: C₃H₅NO₄
MDL Number: MFCD00075480
CAS Number: 2483-57-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty