Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Phenylglycinonitrile 95%

Supplier: AMBEED, INC
Warning

A1467927-100G A1467927-25G A1467927-500G
76662-624EA 19 USD InStock
76662-624 76662-626 76662-628
N-Phenylglycinonitrile 95%
N-Phenylglycinonitrile
Precaución: for Research Use Only


Formula: C₈H₈N₂
MDL Number: MFCD00044430
CAS Number: 3009-97-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty