Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Bromo-2,6-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 99% (by HPLC)

Supplier: AMBEED, INC
Warning

A1467991-1G A1467991-250MG A1467991-25G A1467991-5G
76662-636EA 44 USD InStock
76662-636 76662-638 76662-640 76662-642
2-(4-Bromo-2,6-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 99% (by HPLC)
2-(4-Bromo-2,6-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only


Formula: C₁₂H₁₄BBrF₂O₂
MW: 318.95 g/mol
MDL Number: MFCD26143393
CAS Number: 1799485-20-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty