Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-(ethoxymethylene)-4,4,4-trifluoro-3-oxobutyrate 80% + (mixture of isomers)

Supplier: AMBEED, INC
Warning

A1471440-25G A1471440-500G A1471440-5G
76662-882EA 29 USD InStock
76662-882 76662-884 76662-886
Ethyl-2-(ethoxymethylene)-4,4,4-trifluoro-3-oxobutyrate 80% + (mixture of isomers)
Ethyl-2-(ethoxymethylene)-4,4,4-trifluoro-3-oxobutyrate
Precaución: for Research Use Only


Formula: C₉H₁₁F₃O₄
MDL Number: MFCD02677683
CAS Number: 571-55-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty