Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dibromosulfanilamide 97%

Supplier: AMBEED, INC
Warning

A1471522-100G A1471522-25G A1471522-500G
76662-888EA 180 USD InStock
76662-888 76662-890 76662-892
3,5-Dibromosulfanilamide 97%
3,5-Dibromosulphanilamide
Precaución: for Research Use Only


Formula: C₆H₆Br₂N₂O₂S
MDL Number: MFCD00014783
CAS Number: 39150-45-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty