Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,9-Divinyl-2,4,8,10-tetraoxaspiro[5.5]undecane 99%

Supplier: AMBEED, INC

A1472739-100G A1472739-25G A1472739-500G
76662-974EA 23 USD InStock
76662-974 76662-976 76662-978
3,9-Divinyl-2,4,8,10-tetraoxaspiro[5.5]undecane 99%
3,9-Divinyl-2,4,8,10-tetraoxaspiro[5.5]undecane
Precaución: for Research Use Only


Formula: C₁₁H₁₆O₄
MDL Number: MFCD00006564
CAS Number: 78-19-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty