Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(α)-Boc-DL-2,3-diaminopropionic acid 98%

Supplier: AMBEED, INC
Warning

A1473839-100G A1473839-25G A1473839-5G
76663-150EA 275 USD InStock
76663-150 76663-152 76663-154
N-(α)-Boc-DL-2,3-diaminopropionic acid 98%
N-(alpha)-Boc-DL-2,3-diaminopropionic acid
Precaución: for Research Use Only


Formula: C₈H₁₆N₂O₄
MW: 204.23 g/mol
MDL Number: MFCD00736752
CAS Number: 159002-17-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty