Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-D-2,4-diaminobutyric acid 98%

Supplier: AMBEED, INC
Warning

A1473845-1G A1473845-250MG A1473845-5G
76663-168EA 18 USD InStock
76663-168 76663-170 76663-172
Boc-D-2,4-diaminobutyric acid 98%
Boc-D-2,4-diaminobutyric acid
Precaución: for Research Use Only


Formula: C₉H₁₈N₂O₄
MW: 218.253 g/mol
MDL Number: MFCD00798622
CAS Number: 80445-78-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty