Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-Cbz-α-phosphonoglycine trimethyl ester 98%

Supplier: AMBEED, INC
Warning

A1473853-100G A1473853-25G A1473853-500G A1473853-5G
76663-180EA 71 USD InStock
76663-180 76663-182 76663-184 76663-186
(±)-Cbz-α-phosphonoglycine trimethyl ester 98%
(±)-Cbz-α-phosphonoglycine trimethyl ester
Precaución: for Research Use Only


Formula: C₁₃H₁₈NO₇P
MW: 331.27 g/mol
MDL Number: MFCD00043304
CAS Number: 88568-95-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty