Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

8-Chloro-3-fluoro-1,5-naphthyridine 98+%

Supplier: AMBEED, INC
Warning

A1474747-100MG A1474747-1G A1474747-250MG
76663-248EA 20 USD InStock
76663-248 76663-250 76663-252
8-Chloro-3-fluoro-1,5-naphthyridine 98+%
8-Chloro-3-fluoro-1,5-naphthyridine
Precaución: for Research Use Only


Formula: C₈H₄ClFN₂
MDL Number: MFCD28098174
CAS Number: 1151802-02-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty