Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-5-fluoroisonicotinic acid 99%

Supplier: AMBEED, INC
Warning

A1475531-100G A1475531-1G A1475531-25G A1475531-5G
76663-328EA 420 USD InStock
76663-328 76663-330 76663-332 76663-334
2-Bromo-5-fluoroisonicotinic acid 99%
2-Bromo-5-fluoroisonicotinic acid
Precaución: for Research Use Only


Formula: C₆H₃BrFNO₂
MW: 219.9959 g/mol
MDL Number: MFCD09261246
CAS Number: 885588-12-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty