Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-4-(trifluoromethyl)benzonitrile 98%

Supplier: AMBEED, INC
Warning

A1475737-100G A1475737-1G A1475737-25G A1475737-5G
76663-352EA 339 USD InStock
76663-352 76663-354 76663-356 76663-358
3-Chloro-4-(trifluoromethyl)benzonitrile 98%
3-Chloro-4-(trifluoromethyl)benzonitrile
Precaución: for Research Use Only


Formula: C₈H₃ClF₃N
MW: 205.57
MDL Number: MFCD04972703
CAS Number: 1092460-79-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty