Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-3-(trifluoromethyl)benzonitrile 99+%

Supplier: AMBEED, INC
Warning

A1475815-100G A1475815-25G A1475815-500G A1475815-5G
76663-376EA 117 USD InStock
76663-376 76663-378 76663-380 76663-382
4-Fluoro-3-(trifluoromethyl)benzonitrile 99+%
4-Fluoro-3-(trifluoromethyl)benzonitrile
Precaución: for Research Use Only


Formula: C₈H₃F₄N
MW: 189.11 g/mol
MDL Number: MFCD00061284
CAS Number: 67515-59-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty