Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 3-chloro-2-fluorobenzoate 99+%

Supplier: AMBEED, INC
Warning

A1475845-100G A1475845-25G A1475845-5G
76663-410EA 66 USD InStock
76663-410 76663-412 76663-414
Ethyl 3-chloro-2-fluorobenzoate 99+%
Ethyl 3-chloro-2-fluorobenzoate
Precaución: for Research Use Only


Formula: C₉H₈ClFO₂
MDL Number: MFCD06204525
CAS Number: 773135-55-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty