Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-4-fluorophenylacetonitrile 99+%

Supplier: AMBEED, INC
Warning

A1475857-100G A1475857-1G A1475857-25G A1475857-5G
76663-416EA 292 USD InStock
76663-416 76663-418 76663-420 76663-422
3-Chloro-4-fluorophenylacetonitrile 99+%
3-Chloro-4-fluorophenylacetonitrile
Precaución: for Research Use Only


Formula: C₈H₅ClFN
MDL Number: MFCD01631553
CAS Number: 658-98-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty