Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-chloro-4-fluorobenzoate 99+%

Supplier: AMBEED, INC
Warning

A1475865-100G A1475865-25G A1475865-5G
76663-436EA 83 USD InStock
76663-436 76663-438 76663-440
Methyl-3-chloro-4-fluorobenzoate 99+%
Methyl-3-chloro-4-fluorobenzoate
Precaución: for Research Use Only


Formula: C₈H₆ClFO₂
MW: 188.59 g/mol
MDL Number: MFCD06203675
CAS Number: 234082-35-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty