Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2,3-difluoroanisole 99+%

Supplier: AMBEED, INC
Warning

A1475869-100G A1475869-1G A1475869-25G A1475869-5G
76663-452EA 274 USD InStock
76663-452 76663-454 76663-456 76663-458
4-Bromo-2,3-difluoroanisole 99+%
4-Bromo-2,3-difluoroanisole
Precaución: for Research Use Only


Formula: C₇H₅BrF₂O
MDL Number: MFCD07368017
CAS Number: 406482-22-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty