Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-4-methylanisole 99%

Supplier: AMBEED, INC
Warning

A1475873-100G A1475873-25G A1475873-5G
76663-460EA 153 USD InStock
76663-460 76663-462 76663-464
3-Chloro-4-methylanisole 99%
3-Chloro-4-methylanisole
Precaución: for Research Use Only


Formula: C₈H₉ClO
MDL Number: MFCD00087376
CAS Number: 54788-38-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty