Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxy-3-methylbenzonitrile 99+%

Supplier: AMBEED, INC
Warning

A1475882-100G A1475882-1G A1475882-25G A1475882-5G
76663-478EA 319 USD InStock
76663-478 76663-480 76663-482 76663-484
4-Methoxy-3-methylbenzonitrile 99+%
4-Methoxy-3-methylbenzonitrile
Precaución: for Research Use Only


Formula: C₉H₉NO
MW: 147.18 g/mol
MDL Number: MFCD12797513
CAS Number: 53078-71-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty