Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2-bromo-4,5-difluorobenzoate 99+%

Supplier: AMBEED, INC
Warning

A1475885-100G A1475885-25G A1475885-5G
76663-486EA 87 USD InStock
76663-486 76663-488 76663-490
Ethyl 2-bromo-4,5-difluorobenzoate 99+%
Ethyl 2-bromo-4,5-difluorobenzoate
Precaución: for Research Use Only


Formula: C₉H₇BrF₂O₂
MW: 265.051 g/mol
MDL Number: MFCD08458105
CAS Number: 144267-97-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty