Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-chloro-4-methoxybenzoate 99+%

Supplier: AMBEED, INC
Warning

A1475896-100G A1475896-1G A1475896-25G A1475896-5G
76663-530EA 319 USD InStock
76663-530 76663-532 76663-534 76663-536
Methyl 2-chloro-4-methoxybenzoate 99+%
Methyl 2-chloro-4-methoxybenzoate
Precaución: for Research Use Only


Formula: C₉H₉ClO₃
MDL Number: MFCD06797822
CAS Number: 104253-45-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty