Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1R,2R)-1,2-Cyclohexanedimethanol 98%

Supplier: AMBEED, INC
Warning

A1476029-100G A1476029-25G A1476029-5G
76663-578EA 99 USD InStock
76663-578 76663-580 76663-582
(1R,2R)-1,2-Cyclohexanedimethanol 98%
(1R,2R)-1,2-Cyclohexanedimethanol
Precaución: for Research Use Only


Formula: C₈H₁₆O₂
MDL Number: MFCD16660860
CAS Number: 65376-05-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty